Assay Details
| Gene Symbol: Mrps35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145573.2 | NP_663548.2 | 3 - 4 | 342 | 64 | |
| GenBank mRNA | AK077677.1 | - | 3 - 4 | 343 | 64 | |
| AK170480.1 | - | 3 - 4 | 342 | 64 | ||
| BC019964.1 | - | 3 - 4 | 320 | 4158657 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mrps35
Genomic Map