Assay Details
| Gene Symbol: Aebp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178803.2 | NP_848918.1 | 3 - 4 | 711 | 64 | |
| GenBank mRNA | AB221647.1 | - | 3 - 4 | 362 | 64 | |
| AF090326.1 | - | 446 | 64 | |||
| AK029011.1 | - | 3 - 4 | 713 | 64 | ||
| AK045838.1 | - | 3 - 4 | 477 | 64 | ||
| AK045987.1 | - | 3 - 4 | 477 | 64 | ||
| AK155272.1 | - | 3 - 4 | 443 | 64 | ||
| BC025467.1 | - | 3 - 4 | 1169 | 5351959 | 64 | |
| BC031376.1 | - | 4 - 5 | 572 | 5356616 | 64 | |
| BC057681.1 | - | 3 - 4 | 491 | 3499299 | 64 | |
More Information
Set Membership: |
|

Aebp2
Genomic Map