Assay Details
| Gene Symbol: Slco1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177772.1 | NP_001171243.1 | 12 - 13 | 2129 | 164 | |
| NM_021471.2 | NP_067446.1 | 12 - 13 | 2129 | 164 | ||
| GenBank mRNA | AK045782.1 | - | 3470 | 164 | ||
| AK048926.1 | - | 12 - 13 | 2099 | 164 | ||
| AK161010.1 | - | 3437 | 164 | |||
| AY007379.1 | - | 12 - 13 | 1965 | 164 | ||
| BC078456.1 | - | 12 - 13 | 2038 | 30360663 | 164 | |
| BE865248.1 | - | 3 - 4 | 392 | 164 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slco1c1
Genomic Map