Assay Details
Gene Symbol: Ppp1r36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163103.1 | NP_001156575.1 | 1 - 2 | 118 | 123 | |
GenBank mRNA | AK133227.1 | - | 1 - 2 | 97 | 123 | |
CK137158.1 | - | 1 - 2 | 118 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
