Assay Details
| Gene Symbol: Ppp1r36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163103.1 | NP_001156575.1 | 2 - 3 | 187 | 108 | |
| GenBank mRNA | AK133227.1 | - | 2 - 3 | 166 | 108 | |
| CK137158.1 | - | 2 - 3 | 187 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r36
Genomic Map