Assay Details
| Gene Symbol: Plbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK170196.1 | - | 1 - 2 | 315 | 162 | |
| AK172021.1 | - | 1 - 2 | 229 | 162 | ||
| BC019793.1 | - | 1 - 2 | 282 | 5133074 | 162 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK172021.1, BC019793.1 transcripts.
Set Membership: |
|

Plbd1
Genomic Map