Assay Details
| Gene Symbol: Plbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025806.2 | NP_080082.1 | 6 - 7 | 1059 | 153 | |
| GenBank mRNA | AK003169.1 | - | 6 - 7 | 981 | 153 | |
| AK170196.1 | - | 6 - 7 | 1044 | 153 | ||
| AK172021.1 | - | 6 - 7 | 959 | 153 | ||
| BC019793.1 | - | 6 - 7 | 1011 | 5133074 | 153 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plbd1
Genomic Map