Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400005577.mm.1 |
NM_001282014.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400005577.mm.1 |
NM_001282015.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400005577.mm.1 |
NM_031202.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400005577.mm.1 |
TC0400000810.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400005572.mm.1 |
NM_001282014.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400005572.mm.1 |
NM_001282015.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400005572.mm.1 |
NM_031202.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400005572.mm.1 |
TC0400000810.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400005574.mm.1 |
NM_001282014.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400005574.mm.1 |
NM_001282015.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400005574.mm.1 |
NM_031202.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400005574.mm.1 |
TC0400000810.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400005578.mm.1 |
NM_001282014.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400005578.mm.1 |
NM_001282015.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400005578.mm.1 |
NM_031202.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400005578.mm.1 |
TC0400000810.mm.1 |
MTA TC |