Assay Details
Gene Symbol: Ttc39b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027238.2 | NP_081514.1 | 6 - 7 | 575 | 62 | |
GenBank mRNA | AK037208.1 | - | 6 - 7 | 576 | 62 | |
AK049108.1 | - | 6 - 7 | 626 | 62 | ||
BC151021.1 | - | 6 - 7 | 537 | 9087934 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
