Assay Details
| Gene Symbol: Rin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028724.4 | NP_083000.5 | 10 - 11 | 2349 | 87 | |
| GenBank mRNA | AK014548.1 | - | 2 - 3 | 821 | 87 | |
| AK040763.1 | - | 2 - 3 | 821 | 87 | ||
| AK150680.1 | - | 8 - 9 | 2157 | 87 | ||
| AK151332.1 | - | 7 - 9 | 2113 | 87 | ||
| AK152025.1 | - | 2 - 4 | 794 | 87 | ||
| AK152893.1 | - | 8 - 9 | 2157 | 87 | ||
| AK154421.1 | - | 2 - 3 | 765 | 87 | ||
| AK163047.1 | - | 2 - 3 | 1336 | 87 | ||
| AK171064.1 | - | 2 - 3 | 736 | 87 | ||
| AK198881.1 | - | 1 - 2 | 136 | 87 | ||
| BC005529.1 | - | 2 - 3 | 782 | 3495681 | 87 | |
| BC040390.1 | - | 10 - 11 | 2343 | 3156682 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rin2
Genomic Map