Assay Details
| Gene Symbol: Nop56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024193.2 | NP_077155.2 | 1 - 2 | 63 | 75 | |
| GenBank mRNA | CJ162437.1 | - | 1 - 2 | 66 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_024193.2 transcripts.
Set Membership: |
|

Nop56
Genomic Map