Assay Details
| Gene Symbol: Sirpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177647.2 | NP_001171118.1 | 4 - 5 | 1022 | 84 | |
| NM_001291019.1 | NP_001277948.1 | 6 - 7 | 1676 | 84 | ||
| NM_001291020.1 | NP_001277949.1 | 7 - 8 | 1478 | 84 | ||
| NM_001291021.1 | NP_001277950.1 | 4 - 5 | 1022 | 84 | ||
| NM_001291022.1 | NP_001277951.1 | 3 - 4 | 665 | 84 | ||
| NM_007547.4 | NP_031573.2 | 6 - 7 | 1676 | 84 | ||
| GenBank mRNA | AB018194.1 | - | 6 - 7 | 1242 | 84 | |
| AF072543.1 | - | 6 - 7 | 1232 | 84 | ||
| AF072544.1 | - | 4 - 5 | 578 | 84 | ||
| AF332079.1 | - | 6 - 7 | 1242 | 84 | ||
| AF332080.1 | - | 6 - 7 | 1242 | 84 | ||
| AK159617.1 | - | 7 - 8 | 1485 | 84 | ||
| BC062197.1 | - | 6 - 7 | 1267 | 5368250 | 84 | |
| CJ151068.1 | - | 3 - 4 | 266 | 84 | ||
| CX236057.1 | - | 4 - 5 | 607 | 84 | ||
| D87967.1 | - | 6 - 7 | 1641 | 84 | ||
| D87968.1 | - | 6 - 7 | 1641 | 84 | ||
| Y10349.1 | - | 6 - 7 | 1287 | 84 | ||
More Information
Set Membership: |
|

Sirpa
Genomic Map