Assay Details
| Gene Symbol: Sec23b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252543.1 | NP_001239472.1 | 8 - 9 | 1110 | 75 | |
| NM_001252544.1 | NP_001239473.1 | 8 - 9 | 1103 | 75 | ||
| NM_001252545.1 | NP_001239474.1 | 8 - 9 | 1092 | 75 | ||
| NM_001281816.1 | NP_001268745.1 | 5 - 6 | 732 | 75 | ||
| NM_019787.4 | NP_062761.2 | 8 - 9 | 1121 | 75 | ||
| GenBank mRNA | AF200326.1 | - | 7 - 8 | 993 | 75 | |
| AK014597.1 | - | 8 - 9 | 1071 | 75 | ||
| AK029661.1 | - | 5 - 6 | 733 | 75 | ||
| AK139729.1 | - | 8 - 9 | 1036 | 75 | ||
| AK146083.1 | - | 8 - 9 | 1070 | 75 | ||
| AK149693.1 | - | 8 - 9 | 1050 | 75 | ||
| AK153214.1 | - | 8 - 9 | 1077 | 75 | ||
| AK158398.1 | - | 8 - 9 | 1088 | 75 | ||
| AK167382.1 | - | 8 - 9 | 1059 | 75 | ||
| AK167732.1 | - | 8 - 9 | 1039 | 75 | ||
| AK171597.1 | - | 8 - 9 | 1051 | 75 | ||
| AK209568.1 | - | 1 - 2 | 115 | 75 | ||
| AK220080.1 | - | 1 - 2 | 114 | 75 | ||
| BU614902.1 | - | 4 - 5 | 450 | 75 | ||
| CF724836.1 | - | 2 - 3 | 300 | 75 | ||
More Information
Set Membership: |
|

Sec23b
Genomic Map