Assay Details
Gene Symbol: Crisp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204071.1 | NP_001191000.1 | 5 - 6 | 432 | 67 | |
NM_009420.2 | NP_033446.1 | 5 - 6 | 450 | 67 | ||
GenBank mRNA | AK018856.1 | - | 5 - 6 | 381 | 67 | |
BC049615.1 | - | 5 - 6 | 402 | 6742033 | 67 | |
CJ046590.1 | - | 5 - 6 | 373 | 67 | ||
M25533.1 | - | 5 - 6 | 402 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
