Assay Details
Gene Symbol: Crisp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204071.1 | NP_001191000.1 | 7 - 8 | 657 | 150 | |
NM_009420.2 | NP_033446.1 | 7 - 8 | 675 | 150 | ||
GenBank mRNA | BC049615.1 | - | 7 - 8 | 627 | 6742033 | 150 |
M25533.1 | - | 7 - 8 | 627 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
