Assay Details
| Gene Symbol: Ctdspl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290992.1 | NP_001277921.1 | 3 - 4 | 958 | 135 | |
| NM_212450.4 | NP_997615.1 | 3 - 4 | 962 | 135 | ||
| GenBank mRNA | AK030085.1 | - | 3 - 4 | 478 | 135 | |
| AK033382.1 | - | 3 - 4 | 511 | 135 | ||
| AK041422.1 | - | 3 - 4 | 478 | 135 | ||
| AK044805.1 | - | 3 - 4 | 492 | 135 | ||
| AK049463.1 | - | 3 - 4 | 478 | 135 | ||
| AK082646.1 | - | 3 - 4 | 818 | 135 | ||
| AK160431.1 | - | 3 - 4 | 477 | 135 | ||
| AK163262.1 | - | 3 - 4 | 963 | 135 | ||
| BC052660.1 | - | 3 - 4 | 30049400 | 135 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001290992.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ctdspl2
Genomic Map