Assay Details
| Gene Symbol: Ctdspl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290991.1 | NP_001277920.1 | 5 - 6 | 1329 | 80 | |
| NM_001290992.1 | NP_001277921.1 | 5 - 6 | 1328 | 80 | ||
| NM_212450.4 | NP_997615.1 | 5 - 6 | 1332 | 80 | ||
| GenBank mRNA | AK028251.1 | - | 3 - 4 | 392 | 80 | |
| AK030085.1 | - | 5 - 6 | 848 | 80 | ||
| AK033382.1 | - | 5 - 6 | 881 | 80 | ||
| AK041422.1 | - | 5 - 6 | 848 | 80 | ||
| AK044805.1 | - | 5 - 6 | 863 | 80 | ||
| AK049463.1 | - | 5 - 6 | 848 | 80 | ||
| AK053513.1 | - | 5 - 6 | 858 | 80 | ||
| AK082646.1 | - | 5 - 6 | 1188 | 80 | ||
| AK160431.1 | - | 5 - 6 | 847 | 80 | ||
| AK163262.1 | - | 5 - 6 | 1333 | 80 | ||
| AK169248.1 | - | 4 - 5 | 724 | 80 | ||
| BC052660.1 | - | 5 - 6 | 815 | 30049400 | 80 | |
| BC066219.1 | - | 5 - 6 | 849 | 80 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ctdspl2
Genomic Map