Assay Details
Gene Symbol: Ctdspl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290991.1 | NP_001277920.1 | 6 - 7 | 1399 | 70 | |
NM_001290992.1 | NP_001277921.1 | 6 - 7 | 1398 | 70 | ||
NM_212450.4 | NP_997615.1 | 6 - 7 | 1402 | 70 | ||
GenBank mRNA | AK028251.1 | - | 4 - 5 | 462 | 70 | |
AK030085.1 | - | 6 - 7 | 918 | 70 | ||
AK033382.1 | - | 6 - 7 | 951 | 70 | ||
AK044805.1 | - | 6 - 7 | 933 | 70 | ||
AK049463.1 | - | 6 - 7 | 918 | 70 | ||
AK053513.1 | - | 6 - 7 | 928 | 70 | ||
AK082646.1 | - | 6 - 7 | 1258 | 70 | ||
AK160431.1 | - | 6 - 7 | 917 | 70 | ||
AK163262.1 | - | 6 - 7 | 1403 | 70 | ||
AK169248.1 | - | 5 - 6 | 794 | 70 | ||
BC052660.1 | - | 6 - 7 | 885 | 30049400 | 70 | |
BC066219.1 | - | 6 - 7 | 919 | 70 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
