Assay Details
Gene Symbol: Nlrp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002894.2 | NP_001002894.2 | 2 - 3 | 147 | ||
GenBank mRNA | AY673647.1 | - | 2 - 3 | 147 | ||
BC141199.1 | - | 2 - 3 | 9055811 | 147 | ||
CJ053047.1 | - | 2 - 3 | 147 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
