Assay Details
Gene Symbol: Nlrp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002894.2 | NP_001002894.2 | 6 - 7 | 2274 | 109 | |
GenBank mRNA | AK014932.1 | - | 2 - 3 | 432 | 109 | |
AY596199.1 | - | 5 - 6 | 2150 | 109 | ||
AY673647.1 | - | 6 - 7 | 2274 | 109 | ||
BC141199.1 | - | 6 - 7 | 2258 | 9055811 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
