Assay Details
| Gene Symbol: Gltscr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100452.1 | NP_001093922.1 | 9 - 10 | 2769 | 92 | |
| GenBank mRNA | AB093219.1 | - | 6 - 7 | 1922 | 92 | |
| AK029927.1 | - | 10 - 11 | 93 | |||
| BC032203.1 | - | 6 - 7 | 1450 | 92 | ||
| BC151032.1 | - | 9 - 10 | 2769 | 9087945 | 92 | |
| BC151054.1 | - | 9 - 10 | 2769 | 9087967 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gltscr1l
Genomic Map