Assay Details
| Gene Symbol: Gltscr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100452.1 | NP_001093922.1 | 3 - 4 | 583 | 80 | |
| GenBank mRNA | AK029927.1 | - | 4 - 5 | 227 | 80 | |
| BC151032.1 | - | 3 - 4 | 583 | 9087945 | 80 | |
| BC151054.1 | - | 3 - 4 | 583 | 9087967 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gltscr1l
Genomic Map