Assay Details
| Gene Symbol: Gltscr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100452.1 | NP_001093922.1 | 6 - 7 | 2458 | 74 | |
| GenBank mRNA | AB093219.1 | - | 3 - 4 | 1611 | 74 | |
| AK029927.1 | - | 7 - 8 | 2102 | 74 | ||
| BC032203.1 | - | 3 - 4 | 1139 | 74 | ||
| BC151032.1 | - | 6 - 7 | 2458 | 9087945 | 74 | |
| BC151054.1 | - | 6 - 7 | 2458 | 9087967 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gltscr1l
Genomic Map