Assay Details
| Gene Symbol: Chchd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025366.3 | NP_079642.1 | 1 - 2 | 174 | 67 | |
| GenBank mRNA | AK003263.1 | - | 1 - 2 | 133 | 67 | |
| AK010311.1 | - | 1 - 2 | 117 | 67 | ||
| AK011261.1 | - | 1 - 2 | 132 | 67 | ||
| BC052720.1 | - | 1 - 2 | 153 | 6818266 | 67 | |
| BC064310.1 | - | 1 - 2 | 153 | 67 | ||
| BC092216.1 | - | 1 - 2 | 141 | 30609530 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chchd1
Genomic Map