Assay Details
Gene Symbol: Parva | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020606.5 | NP_065631.3 | 3 - 4 | 563 | 82 | |
GenBank mRNA | AB045321.1 | - | 3 - 4 | 436 | 82 | |
AF237774.1 | - | 3 - 4 | 436 | 82 | ||
AF264766.1 | - | 3 - 4 | 333 | 82 | ||
AK077368.1 | - | 4 - 5 | 395 | 82 | ||
AK148878.1 | - | 3 - 4 | 297 | 82 | ||
AK162796.1 | - | 4 - 5 | 458 | 82 | ||
BC039750.1 | - | 3 - 4 | 321 | 82 | ||
BC059236.1 | - | 3 - 4 | 563 | 6400393 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
