Assay Details
| Gene Symbol: Zeb2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110572.1 | 1 - 2 | 167 | 71 | ||
| GenBank mRNA | BC031611.1 | - | 1 - 2 | 167 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zeb2os| Gene Symbol: Zeb2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110572.1 | 1 - 2 | 167 | 71 | ||
| GenBank mRNA | BC031611.1 | - | 1 - 2 | 167 | 71 | |
Set Membership: |
|