Assay Details
| Gene Symbol: Itgb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159564.1 | NP_001153036.1 | 16 - 17 | 2438 | 94 | |
| NM_021359.3 | NP_067334.1 | 13 - 14 | 2332 | 94 | ||
| GenBank mRNA | AF115376.1 | - | 16 - 17 | 2284 | 94 | |
| AK036439.1 | - | 16 - 17 | 2439 | 94 | ||
| AK085900.1 | - | 16 - 17 | 2479 | 94 | ||
| BC049185.1 | - | 13 - 14 | 2301 | 6475423 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Itgb6
Genomic Map