Assay Details
| Gene Symbol: Slc35b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF414190.1 | - | 1 - 2 | 108 | 53 | |
| AK003377.1 | - | 1 - 2 | 117 | 53 | ||
| AK144403.1 | - | 1 - 2 | 131 | 53 | ||
| AK152362.1 | - | 1 - 2 | 109 | 53 | ||
| AK154868.1 | - | 1 - 2 | 94 | 53 | ||
| AK155551.1 | - | 1 - 2 | 141 | 53 | ||
| BC025875.1 | - | 1 - 2 | 107 | 5137689 | 53 | |
| BC036992.1 | - | 1 - 2 | 115 | 5003855 | 53 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF414190.1, AK003377.1, AK144403.1, AK152362.1, AK154868.1, AK155551.1, BC025875.1, BC036992.1 transcripts.
Set Membership: |
|

Slc35b2
Genomic Map