Assay Details
| Gene Symbol: Slc35b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028662.2 | NP_082938.1 | 1 - 2 | 624 | 82 | |
| GenBank mRNA | AF414190.1 | - | 2 - 3 | 297 | 82 | |
| AK003377.1 | - | 2 - 3 | 306 | 82 | ||
| AK144403.1 | - | 2 - 3 | 320 | 82 | ||
| AK152362.1 | - | 2 - 3 | 298 | 82 | ||
| AK154868.1 | - | 2 - 3 | 283 | 82 | ||
| AK155551.1 | - | 2 - 3 | 330 | 82 | ||
| AK167343.1 | - | 1 - 2 | 625 | 82 | ||
| BC025875.1 | - | 2 - 3 | 296 | 5137689 | 82 | |
| BC036992.1 | - | 2 - 3 | 304 | 5003855 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc35b2
Genomic Map