Assay Details
Gene Symbol: Palb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081238.2 | NP_001074707.1 | 9 - 10 | 2930 | 86 | |
NM_001289842.1 | NP_001276771.1 | 9 - 10 | 2930 | 86 | ||
NM_001289843.1 | NP_001276772.1 | 8 - 9 | 2836 | 86 | ||
NM_001289844.1 | NP_001276773.1 | 9 - 10 | 1841 | 86 | ||
NM_001289845.1 | NP_001276774.1 | 6 - 7 | 770 | 86 | ||
GenBank mRNA | AK154227.1 | - | 9 - 10 | 2902 | 86 | |
AK156701.1 | - | 9 - 10 | 2849 | 86 | ||
AK157382.1 | - | 9 - 10 | 2849 | 86 | ||
BB757034.1 | - | 1 - 2 | 121 | 86 | ||
BC044748.2 | - | 5 - 6 | 535 | 86 | ||
BC055302.1 | - | 6 - 7 | 733 | 86 | ||
BC066139.1 | - | 8 - 9 | 2778 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
