Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700035194.mm.1 |
NM_001289842.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035194.mm.1 |
NM_001289843.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035194.mm.1 |
TC0700004225.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700035197.mm.1 |
NM_001081238.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035197.mm.1 |
NM_001289842.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035197.mm.1 |
NM_001289843.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035197.mm.1 |
NM_001289844.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035197.mm.1 |
NM_001289845.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035197.mm.1 |
TC0700004225.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700035195.mm.1 |
NM_001081238.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035195.mm.1 |
NM_001289842.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035195.mm.1 |
NM_001289843.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035195.mm.1 |
NM_001289844.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035195.mm.1 |
NM_001289845.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035195.mm.1 |
TC0700004225.mm.1 |
MTA TC |