| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200009506.mm.1 |
NM_175513.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200009506.mm.1 |
NM_175513.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200009506.mm.1 |
TC0200001189.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200009508.mm.1 |
NM_175513.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200009508.mm.1 |
NM_175513.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200009508.mm.1 |
TC0200001189.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200009509.mm.1 |
NM_175513.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200009509.mm.1 |
NM_175513.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200009509.mm.1 |
TC0200001189.mm.1 |
MTA TC |