Assay Details
| Gene Symbol: Edrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178115.4 | NP_835216.3 | 1 - 2 | 247 | 115 | |
| GenBank mRNA | AK052387.1 | - | 1 - 2 | 230 | 115 | |
| AK144179.1 | - | 1 - 2 | 208 | 115 | ||
| AK144959.1 | - | 1 - 2 | 217 | 115 | ||
| BC072596.1 | - | 1 - 2 | 216 | 6409214 | 115 | |
| CJ078287.1 | - | 1 - 2 | 251 | 115 | ||
More Information
Set Membership: |
|

Edrf1
Genomic Map