Assay Details
Gene Symbol: Fermt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198029.2 | NP_932146.2 | 10 - 11 | 1995 | 67 | |
GenBank mRNA | AF073489.1 | - | 7 - 8 | 781 | 67 | |
AK030947.1 | - | 9 - 10 | 2185 | 67 | ||
AK050804.1 | - | 9 - 10 | 1143 | 67 | ||
BC029093.1 | - | 1 - 2 | 114 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC029093.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
