Assay Details
| Gene Symbol: Jag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013822.5 | NP_038850.1 | 20 - 21 | 2717 | 76 | |
| GenBank mRNA | AF171092.1 | - | 20 - 21 | 2722 | 76 | |
| AK137092.1 | - | 20 - 21 | 2617 | 76 | ||
| AK162228.1 | - | 3 - 4 | 219 | 76 | ||
| BC058675.1 | - | 20 - 21 | 2529 | 6834418 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Jag1
Genomic Map