Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200000967.mm.1 |
NM_001146119.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000967.mm.1 |
NM_001146119.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000967.mm.1 |
NM_029758.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000967.mm.1 |
NM_029758.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000967.mm.1 |
TC1200000098.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200000968.mm.1 |
NM_001146119.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000968.mm.1 |
NM_001146119.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000968.mm.1 |
NM_029758.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000968.mm.1 |
NM_029758.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000968.mm.1 |
TC1200000098.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200000969.mm.1 |
NM_001146119.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000969.mm.1 |
NM_001146119.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000969.mm.1 |
NM_029758.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000969.mm.1 |
NM_029758.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000969.mm.1 |
TC1200000098.mm.1 |
MTA TC |