Assay Details
| Gene Symbol: Laptm4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008640.2 | NP_032666.2 | 3 - 4 | 1192 | 120 | |
| GenBank mRNA | AK013963.1 | - | 2 - 3 | 210 | 120 | |
| AK075699.1 | - | 3 - 4 | 427 | 120 | ||
| AK084515.1 | - | 3 - 4 | 835 | 120 | ||
| AK088958.1 | - | 3 - 4 | 452 | 120 | ||
| AK165638.1 | - | 3 - 4 | 461 | 120 | ||
| BC086667.1 | - | 3 - 4 | 372 | 30633914 | 120 | |
| BC089021.1 | - | 3 - 4 | 405 | 6734038 | 120 | |
| U34259.1 | - | 4 - 5 | 1059 | 120 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Laptm4a
Genomic Map