Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200007556.mm.1 |
NM_001099298.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007556.mm.1 |
TC0200000953.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200007565.mm.1 |
NM_001099298.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007565.mm.1 |
TC0200000953.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200007566.mm.1 |
NM_001099298.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007566.mm.1 |
TC0200000953.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200007554.mm.1 |
NM_001099298.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007554.mm.1 |
TC0200000953.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200007560.mm.1 |
NM_001099298.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007560.mm.1 |
TC0200000953.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200007552.mm.1 |
NM_001099298.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007552.mm.1 |
TC0200000953.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200007563.mm.1 |
NM_001099298.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007563.mm.1 |
TC0200000953.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200007557.mm.1 |
NM_001099298.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007557.mm.1 |
TC0200000953.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200007558.mm.1 |
NM_001099298.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007558.mm.1 |
TC0200000953.mm.1 |
MTA TC |