Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300015437.mm.1 |
NM_001081262.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015437.mm.1 |
NM_001081262.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015437.mm.1 |
TC0300002043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300015439.mm.1 |
NM_001081262.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015439.mm.1 |
NM_001081262.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015439.mm.1 |
TC0300002043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300015444.mm.1 |
NM_001081262.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015444.mm.1 |
NM_001081262.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015444.mm.1 |
TC0300002043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300015448.mm.1 |
NM_001081262.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015448.mm.1 |
NM_001081262.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015448.mm.1 |
TC0300002043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300015449.mm.1 |
NM_001081262.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015449.mm.1 |
NM_001081262.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015449.mm.1 |
TC0300002043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300015438.mm.1 |
NM_001081262.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015438.mm.1 |
NM_001081262.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015438.mm.1 |
TC0300002043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300015447.mm.1 |
NM_001081262.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015447.mm.1 |
NM_001081262.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015447.mm.1 |
TC0300002043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300015451.mm.1 |
NM_001081262.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015451.mm.1 |
NM_001081262.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015451.mm.1 |
TC0300002043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300015442.mm.1 |
NM_001081262.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015442.mm.1 |
NM_001081262.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015442.mm.1 |
TC0300002043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300015446.mm.1 |
NM_001081262.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015446.mm.1 |
NM_001081262.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015446.mm.1 |
TC0300002043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300015450.mm.1 |
NM_001081262.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015450.mm.1 |
NM_001081262.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015450.mm.1 |
TC0300002043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300015445.mm.1 |
NM_001081262.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015445.mm.1 |
NM_001081262.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015445.mm.1 |
TC0300002043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300015440.mm.1 |
NM_001081262.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015440.mm.1 |
NM_001081262.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015440.mm.1 |
TC0300002043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300015441.mm.1 |
NM_001081262.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015441.mm.1 |
NM_001081262.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015441.mm.1 |
TC0300002043.mm.1 |
MTA TC |