Assay Details
Gene Symbol: Erich6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081262.1 | NP_001074731.1 | 13 - 14 | 1923 | 62 | |
GenBank mRNA | AK133299.1 | - | 10 - 11 | 2525 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1961928 | AK133299.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1961928 | NM_001081262.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 809447 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 96004 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 12723 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 82536 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 604983 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 786355 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 53676 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902101 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 329248 | AK007542.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 329248 | AK009455.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 329248 | AK144593.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 329248 | BC023111.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 329248 | BC039626.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 329248 | BC062173.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 329248 | BC089556.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 329248 | BC099489.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 329248 | NM_028000.1 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 60025 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 655828 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 802884 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 418674 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 44277 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 90876 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 37121 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1961928 | AK133299.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1961928 | NM_001081262.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00651541 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04920190 | - | - |
Whole Mouse Genome G4122A | A_52_P226466 | AK133299.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P226466 | NM_001081262.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04920190 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169196 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169402 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169191 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169381 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169354 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169340 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169345 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169187 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169194 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169359 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169282 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169329 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169375 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169240 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169234 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169308 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169343 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169268 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169370 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169387 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169214 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169256 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169333 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169305 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169208 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169307 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169372 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169289 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169203 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169384 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169177 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169227 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169292 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169312 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169225 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169367 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169273 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169331 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169204 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169207 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169222 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169291 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169337 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169245 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169262 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169193 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169199 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169364 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169362 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169322 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169212 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169356 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169317 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169215 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169260 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169294 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169287 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169192 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169298 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169272 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169377 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169202 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169182 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169391 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169335 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169237 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169198 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169205 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169197 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169270 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169242 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169179 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169346 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169266 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169394 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169302 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169180 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169248 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22169210 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169375 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169374 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169342 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169289 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02701184 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169238 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169313 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169298 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169337 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02984912 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02877744 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169362 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169372 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169329 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169310 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169304 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169331 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169351 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02965524 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169322 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169369 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02766079 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02429983 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02701186 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169377 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169326 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169357 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02509800 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02984915 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03060591 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169393 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169242 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02993735 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169381 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169391 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02489553 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02509811 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169340 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169343 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169354 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02485402 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02984913 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169324 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169333 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169240 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169380 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169321 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02946645 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169305 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02984895 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169292 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169332 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02509799 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169308 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169307 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02993731 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169286 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02423014 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169294 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169311 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169237 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02452081 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169346 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169370 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22169312 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10498330 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10498329 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10498325 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10498331 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10498326 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10498324 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1458320_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10498323 | - | - |
Mouse Genome 430 2.0 Array | 1458320_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1458320_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 545527_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 545527_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0300015436.mm.1 | NM_001081262.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300015436.mm.1 | NM_001081262.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300015436.mm.1 | TC0300002043.mm.1 | MTA TC |
Mouse Expression Set 430 | 1458320_at | - | - |
Mouse Expression Set 430 | 1458320_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1458320_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1458320_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 536705 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseRef-8 v2 Expression BeadChip | ILMN_3098994 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3026210 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3098994 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3026210 | - | - |
