Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200010295.mm.1 |
NM_001161111.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200010295.mm.1 |
NM_001161111.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200010295.mm.1 |
NM_172574.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200010295.mm.1 |
NM_172574.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200010295.mm.1 |
TC1200001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200010296.mm.1 |
NM_001161111.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200010296.mm.1 |
NM_001161111.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200010296.mm.1 |
NM_172574.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200010296.mm.1 |
NM_172574.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200010296.mm.1 |
TC1200001464.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200010300.mm.1 |
NM_001161111.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200010300.mm.1 |
NM_001161111.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200010300.mm.1 |
NM_172574.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200010300.mm.1 |
NM_172574.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200010300.mm.1 |
TC1200001464.mm.1 |
MTA TC |