Assay Details
| Gene Symbol: Pdia6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027959.3 | NP_082235.1 | 6 - 7 | 628 | 79 | |
| GenBank mRNA | AK005989.1 | - | 6 - 7 | 634 | 79 | |
| AK076558.1 | - | 6 - 7 | 629 | 79 | ||
| AK133792.1 | - | 6 - 7 | 635 | 79 | ||
| AK152960.1 | - | 6 - 7 | 641 | 79 | ||
| AK165877.1 | - | 6 - 7 | 607 | 79 | ||
| AK166111.1 | - | 6 - 7 | 635 | 79 | ||
| AK167386.1 | - | 6 - 7 | 646 | 79 | ||
| AK168136.1 | - | 6 - 7 | 635 | 79 | ||
| AK168279.1 | - | 6 - 7 | 635 | 79 | ||
| AK211058.1 | - | 1 - 2 | 54 | 79 | ||
| AK217964.1 | - | 1 - 2 | 54 | 79 | ||
| BC006865.1 | - | 6 - 7 | 617 | 2645183 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdia6
Genomic Map