| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200010305.mm.1 |
NM_001201332.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010305.mm.1 |
NM_001201332.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010305.mm.1 |
TC1200001465.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200010303.mm.1 |
NM_001201332.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010303.mm.1 |
NM_001201332.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010303.mm.1 |
NM_001201333.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010303.mm.1 |
NM_001201333.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010303.mm.1 |
TC1200001465.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200010304.mm.1 |
NM_001201332.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010304.mm.1 |
NM_001201332.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010304.mm.1 |
NM_001201333.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010304.mm.1 |
NM_001201333.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010304.mm.1 |
TC1200001465.mm.1 |
MTA TC |