Assay Details
| Gene Symbol: Ebf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110513.1 | NP_001103983.1 | 9 - 10 | 964 | 66 | |
| GenBank mRNA | AF387630.1 | - | 9 - 10 | 964 | 66 | |
| AF387631.1 | - | 9 - 10 | 964 | 66 | ||
| AF387632.1 | - | 9 - 10 | 964 | 66 | ||
| AF387633.1 | - | 9 - 10 | 964 | 66 | ||
| AF387634.1 | - | 9 - 10 | 964 | 66 | ||
| BC138119.1 | - | 9 - 10 | 964 | 8861138 | 66 | |
| BC145155.1 | - | 10 - 11 | 1050 | 9053706 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ebf4
Genomic Map