Assay Details
Gene Symbol: H2-Ab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207105.3 | NP_996988.2 | 2 - 3 | 457 | 112 | |
GenBank mRNA | AF065912.1 | - | 2 - 3 | 360 | 112 | |
AF065913.1 | - | 2 - 3 | 387 | 112 | ||
AF119251.1 | - | 2 - 3 | 366 | 112 | ||
AF119252.1 | - | 2 - 3 | 366 | 112 | ||
AK077436.1 | - | 2 - 3 | 445 | 112 | ||
AK170370.1 | - | 2 - 3 | 439 | 112 | ||
AY303784.1 | - | 1 - 2 | 290 | 112 | ||
AY303785.1 | - | 1 - 2 | 281 | 112 | ||
BC057998.1 | - | 2 - 3 | 415 | 5340289 | 112 | |
M13537.1 | - | 2 - 3 | 112 | |||
M13540.1 | - | 2 - 3 | 366 | 112 | ||
M13541.1 | - | 2 - 3 | 334 | 112 | ||
M66213.1 | - | 2 - 3 | 366 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
