Assay Details
Gene Symbol: H2-Ab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207105.3 | NP_996988.2 | 3 - 4 | 734 | 56 | |
GenBank mRNA | AK077436.1 | - | 3 - 4 | 722 | 56 | |
AK170370.1 | - | 3 - 4 | 716 | 56 | ||
CK330363.1 | - | 3 - 4 | 56 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
