Assay Details
| Gene Symbol: Tmem62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175285.3 | NP_780494.1 | 8 - 9 | 1252 | 104 | |
| GenBank mRNA | AK046793.1 | - | 8 - 9 | 1051 | 104 | |
| BC049167.1 | - | 7 - 8 | 904 | 104 | ||
| BC070410.1 | - | 8 - 9 | 1252 | 5708443 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem62
Genomic Map