Assay Details
Gene Symbol: Fbxl19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172748.2 | NP_766336.2 | 3 - 4 | 714 | 93 | |
GenBank mRNA | AK028405.1 | - | 3 - 4 | 483 | 93 | |
AK031431.1 | - | 2 - 3 | 455 | 93 | ||
BC059812.1 | - | 3 - 4 | 714 | 6401846 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
