Assay Details
| Gene Symbol: Klhl41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081087.1 | NP_001074556.1 | 4 - 5 | 1645 | 117 | |
| GenBank mRNA | AK132622.1 | - | 4 - 5 | 658 | 117 | |
| BC150698.1 | - | 4 - 5 | 1730 | 9087609 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein ubiquitination
myofibril assembly
regulation of lateral pseudopodium assembly
skeletal muscle cell differentiation
sarcomere organization
regulation of myoblast differentiation
skeletal muscle fiber development
regulation of myoblast proliferation
regulation of skeletal muscle cell differentiation
molecular_function
protein ubiquitination
myofibril assembly
regulation of lateral pseudopodium assembly
skeletal muscle cell differentiation
sarcomere organization
regulation of myoblast differentiation
skeletal muscle fiber development
regulation of myoblast proliferation
regulation of skeletal muscle cell differentiation
molecular_function
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P647740 | NM_001081087.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P647740 | AK132622.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P647740 | BC150698.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P647740 | NM_001081087.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P647740 | AK132622.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P647740 | BC150698.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 605402 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 893201 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 631189 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 705046 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 443127 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894824 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 8084 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 76536 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 152182 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P647740 | NM_001081087.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P647740 | AK132622.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P647740 | BC150698.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P647740 | NM_001081087.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P647740 | AK132622.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P647740 | BC150698.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P647740 | NM_001081087.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P647740 | AK132622.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P647740 | BC150698.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P647740 | NM_001081087.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P647740 | AK132622.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P647740 | BC150698.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21310432 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310420 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310438 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310411 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310407 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01206603 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310436 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310450 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01157047 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310444 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01670920 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310434 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310408 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310445 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310417 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310463 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310467 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310449 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310426 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310456 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310464 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310431 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310441 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310432 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310466 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310419 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310423 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01679264 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310412 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01836215 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310446 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310455 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310452 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01157048 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01836217 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01752844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310406 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01679240 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21310462 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10472569 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10472566 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10472563 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10472567 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10472568 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10472564 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10472565 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1444494_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10472562 | - | - |
| Mouse Genome 430 2.0 Array | 1444494_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1444494_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 228003_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 228003_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200007910.mm.1 | NM_001081087.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200007910.mm.1 | NM_001081087.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200007910.mm.1 | TC0200001008.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1444494_PM_at | - | - |
| Mouse Expression Set 430 | 1444494_at | - | - |
| Murine Genome U74v2 Set | 170352_r_at | - | - |
| Murine Genome U74v2 Set | 170352_PM_r_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444494_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444494_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 551372 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2527465 | - | - |

Klhl41
Genomic Map