Assay Details
Gene Symbol: Ssb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110145.1 | NP_001103615.1 | 5 - 6 | 623 | 127 | |
NM_009278.4 | NP_033304.1 | 5 - 6 | 650 | 127 | ||
GenBank mRNA | AK032940.1 | - | 5 - 6 | 567 | 127 | |
AK088650.1 | - | 5 - 6 | 606 | 127 | ||
AK088677.1 | - | 5 - 6 | 557 | 127 | ||
AK134864.1 | - | 5 - 6 | 588 | 127 | ||
AK141487.1 | - | 5 - 6 | 545 | 127 | ||
AK168828.1 | - | 5 - 6 | 573 | 127 | ||
BC003820.1 | - | 5 - 6 | 510 | 3593742 | 127 | |
BC106150.1 | - | 5 - 6 | 503 | 3483153 | 127 | |
L00993.1 | - | 5 - 6 | 586 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
